missing translation for 'onlineSavingsMsg'
Learn More

ML-109, MedChemExpress

Product Code. 16477527
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16477527

Brand: Medchem Express HY114116

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ML-109 is a potent and full thyroid stimulating hormone receptor (TSHR) agonist, with an EC50 of 40 nM.

ML-109 is a potent and full thyroid stimulating hormone receptor (TSHR) agonist, with an EC50 of 40 nM.

Specifications

Chemical Name or Material ML-109
CAS 1186649-91-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H29N3O5
Quantity 5 mg
Solubility Information DMSO : 83.33 mg/mL (159.15 mM; Need ultrasonic)
SMILES CC(NC1=CC=C(OCC2=CC(C(N3CC4=CC=CC=C4)NC5=C(C(O)=CC=C5)C3=O)=CC=C2OC)C=C1)=O
Molecular Weight (g/mol) 523.58
Formula Weight 523.58
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.