missing translation for 'onlineSavingsMsg'
Learn More

MKP-3/DUSP6 Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP2-26174

Product Code. 18197583

  • 506.00 EUR / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MKP-3/DUSP6 Polyclonal specifically detects MKP-3/DUSP6 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MKP-3/DUSP6
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
DUSP6
Peptide with sequence C-PSNQNVYQVDSLQST corresponding to C-Terminus according to NP_073143.2.
0.1 mg
Cell Cycle and Replication
1848
Human, Mouse, Rat, Pig
IgG
Western Blot, ELISA
0.5 mg/mL
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:128000
dual specificity phosphatase 6, Dual specificity protein phosphatase PYST1, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP-3, MKP3serine/threonine specific protein phosphatase, PYST1dual specificity protein phosphatase 6
Goat
Affinity Purified
RUO
Primary
This antibody is expected to recognize both reported isoforms (NP_001937.2; NP_073143.2).
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only