Learn More
MKC9989, MedChemExpress
Product Code. 16485138
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16485138
Brand: Medchem Express HY12399
MedChemExpress MKC9989 is a Hydroxy aryl aldehydes (HAA) inhibitor and also inhibits IRE1α with an IC50 of 0.23 to 44 μM.
MKC9989 is a Hydroxy aryl aldehydes (HAA) inhibitor and also inhibits IRE1α with an IC50 of 0.23 to 44 μM.
Specifications
Chemical Name or Material MKC9989
CAS 1338934-20-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H20O7
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (148.66 mM)
SMILES O=CC1=C(O2)C(C(C)=C(CCOCCOC)C2=O)=CC(OC)=C1O
Molecular Weight (g/mol) 336.34
Formula Weight 336.34
Percent Purity 97.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death