Learn More
MKC8866, MedChemExpress
Product Code. 16492916
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16492916
Brand: Medchem Express HY104040
MedChemExpress MKC8866, a salicylaldehyde analog, is a potent, selective IRE1 RNase inhibitor with an IC50 of 0.29 μM in human vitro.
MKC8866 strongly inhibits Dithiothreitol-induced X-box-binding protein 1-spliced (XBP1s) expression with an EC50 of 0.52 μM and unstresses RPMI 8226 cells with an IC50 of 0.14 μM. MKC8866 inhibits IRE1 RNase in breast cancer cells leading to the decreased production of pro-tumorigenic factors and it can inhibits prostate cancer (PCa) tumor growth.
Specifications
Chemical Name or Material MKC8866
CAS 1338934-59-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H19NO7
Quantity 25 mg
Solubility Information DMSO : 16.67 mg/mL (46.13 mM; Need ultrasonic)
SMILES O=CC1=C(O2)C(C(C)=C(CC(N3CCOCC3)=O)C2=O)=CC(OC)=C1O
Molecular Weight (g/mol) 361.35
Formula Weight 361.35
Percent Purity 98.22%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death