missing translation for 'onlineSavingsMsg'
Learn More

MK6-83, TRC

Product Code. 30478756
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478756 50 mg 50mg
30469186 5 mg 5mg
30477435 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478756 Supplier Toronto Research Chemicals Supplier No. TRCM42460050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1062271-24-2
Molecular Formula C16H20N2O2S2
Molecular Weight (g/mol) 336.47
Synonym MK6-83, 5-Methyl-N-[2-(1-piperidinyl)phenyl]-2-thiophenesulfonamide
IUPAC Name 5-methyl-N-(2-piperidin-1-ylphenyl)thiophene-2-sulfonamide
SMILES Cc1ccc(s1)S(=O)(=O)Nc2ccccc2N3CCCCC3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H20N2O2S2/c1-13-9-10-16(21-13)22(19,20)17-14-7-3-4-8-15(14)18-11-5-2-6-12-18/h3-4,7-10,17H,2,5-6,11-12H2,1H3
Formula Weight 336.0966
Chemical Name or Material 5-Methyl-N-[2-(1-piperidinyl)phenyl]-2-thiophenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.