missing translation for 'onlineSavingsMsg'
Learn More

MK2-IN-1 hydrochloride, MedChemExpress

Product Code. 16498097
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498097 5 mg 5mg
16415388 10 mg 10mg
16401577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498097 Supplier Medchem Express Supplier No. HY12834A5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK2-IN-1 hydrochloride is a potent and selecitve MAPKAPK2(MK2) inhibitor(IC50=0.11 uM) with a non-ATP competitive binding mode.

MK2-IN-1 hydrochloride is a potent and selecitve MAPKAPK2(MK2) inhibitor(IC50=0.11 uM) with a non-ATP competitive binding mode.

Specifications

Chemical Name or Material MK2-IN-1 hydrochloride
CAS 1314118-94-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H26Cl2N4O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (196.30 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C2=CC=C(Cl)C=C2)O1)N(C3=CC=C(N4CCNCC4)C=C3)CC5=NC=CC=C5.[H]Cl
Molecular Weight (g/mol) 509.43
Formula Weight 509.43
Percent Purity 97.32%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.