missing translation for 'onlineSavingsMsg'
Learn More

MK-886, MedChemExpress

Product Code. 16438597
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438597

Brand: Medchem Express HY14166

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-886 (L 663536) is a potent, cell-permeable and orally active FLAP (IC50 of 30 nM) and leukotriene biosynthesis (IC50s of 3 nM and 1.1 μM in intact leukocytes and human whole blood, respectively) inhibitor. MK-886 is also a non-competitive PPARα antagonist and can induce apoptosis.

MK-886 (L 663536) is a potent, cell-permeable and orally active FLAP (IC50 of 30 nM) and leukotriene biosynthesis (IC50s of 3 nM and 1.1 μM in intact leukocytes and human whole blood, respectively) inhibitor. MK-886 is also a non-competitive PPARα antagonist and can induce apoptosis.

Specifications

Chemical Name or Material MK-886
CAS 118414-82-7
Color White
Purity Grade Notes Research
Molecular Formula C27H34ClNO2S
Quantity 5 mg
Synonym L 663536
Solubility Information DMSO : 75 mg/mL (158.87 mM; Need ultrasonic)
SMILES CC(C)C1=CC=C(N(CC2=CC=C(Cl)C=C2)C(CC(C)(C(O)=O)C)=C3SC(C)(C)C)C3=C1
Molecular Weight (g/mol) 472.08
Formula Weight 472.08
Percent Purity 99.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.