missing translation for 'onlineSavingsMsg'
Learn More

MK-5046, MedChemExpress

Product Code. p-200097576
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466446

Brand: Medchem Express HY14342

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-5046 is a novel BRS-3 agonist, binds to BRS-3 with high affinity (mouse Ki = 1.6 nM, human Ki = 25 nM).

MK-5046 is a novel BRS-3 agonist, binds to BRS-3 with high affinity (mouse Ki = 1.6 nM, human Ki = 25 nM).

Specifications

Chemical Name or Material MK-5046
Physical Form Powder
CAS 1022152-70-0
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H18F6N4O
Solubility Information DMSO : 14.29 mg/mL (32.16 mM; Need ultrasonic)
SMILES O[C@](CC1=NC(CC2(C(F)(F)F)CC2)=CN1)(C(F)(F)F)C3=CC=C(N4C=CC=N4)C=C3
Molecular Weight (g/mol) 444.37
Formula Weight 444.37
Percent Purity 99.47%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.