missing translation for 'onlineSavingsMsg'
Learn More

MK-4074, MedChemExpress

Product Code. 16426857
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16426857

missing translation for 'mfr': Medchem Express HY107709

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-4074 is a liver-specific inhibitor of acetyl-CoA carboxylase ACC1 and ACC2 with IC50 values of approximately 3 nM.

MK-4074 is a liver-specific inhibitor of acetyl-CoA carboxylase ACC1 and ACC2 with IC50 values of approximately 3 nM.

Specifications

Chemical Name or Material MK-4074
CAS 1039758-22-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C33H31N3O6
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (88.40 mM; Need ultrasonic)
SMILES O=C1CC2(CCN(C(C3=CC(OC)=C4C(N(C5CC5)C=C4C)=C3)=O)CC2)OC6=CC=C(C7=CN=CC(C(O)=O)=C7)C=C16
Molecular Weight (g/mol) 565.62
Formula Weight 565.62
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.