missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MK-2206 dihydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Description
MK-2206 dihydrochloride (MK-2206 (2HCl)) is an orally active allosteric AKT inhibitor with IC50s of 5 nM, 12 nM, and 65 nM for AKT1, AKT2, and AKT3, respectively. MK-2206 dihydrochloride induces autophagy.
Specifications
Specifications
| Chemical Name or Material | MK-2206 dihydrochloride |
| CAS | 1032350-13-2 |
| Color | Green Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H23Cl2N5O |
| Quantity | 200 mg |
| Synonym | MK-2206 (2HCl) |
| Solubility Information | DMSO : 16.67 mg/mL (34.70 mM; ultrasonic and warming and heat to 70°C) ∣H2O : 3.57 mg/mL (7.43 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C1N2C(C3=CC(C4=CC=CC=C4)=C(N=C3C=C2)C5=CC=C(C6(N)CCC6)C=C5)=NN1.Cl.Cl |
| Molecular Weight (g/mol) | 480.39 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction