Learn More
Macherey-Nagel™ Mix III Diamino Clean-Up Mix
Shop All Macherey-Nagel Products
Click to view available options
Volume (Metric):
15 mL
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Chromabond QuEChERS Clean-up Mixes (EN 15662) |
| Type | Diamino Clean-Up Mix |
| Volume (Metric) | 15 mL |
| Quantity | 50 Pack |