missing translation for 'onlineSavingsMsg'
Learn More

Mivotilate, MedChemExpress

Product Code. 16492868
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492868 10 mg 10mg
16415617 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492868 Supplier Medchem Express Supplier No. HY100242

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mivotilate is a nontoxic, potent activator of the aryl hydrocarbon receptor (AhR), and acts as a hepatoprotective agent.

Mivotilate is a nontoxic, potent activator of the aryl hydrocarbon receptor (AhR), and acts as a hepatoprotective agent.

Specifications

Chemical Name or Material Mivotilate
CAS 130112-42-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H14N2O3S3
Quantity 10 mg
Synonym YH439
Solubility Information DMSO : 12.5 mg/mL (37.83 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(OC(C)C)/C(C(NC1=NC(C)=CS1)=O)=C2SCS\2
Molecular Weight (g/mol) 330.45
Formula Weight 330.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.