Learn More
Mivebresib, MedChemExpress
Product Code. 16499436
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Product Code. 16499436
Brand: Medchem Express HY100015
MedChemExpress Mivebresib (ABBV-075) is a potent and orally active bromodomain and extraterminal domain (BET) bromodomain inhibitor. Mivebresib binds to BRD4 with a Ki of 1.5 nM.
Mivebresib (ABBV-075) is a potent and orally active bromodomain and extraterminal domain (BET) bromodomain inhibitor. Mivebresib binds to BRD4 with a Ki of 1.5 nM.
Specifications
Chemical Name or Material Mivebresib
CAS 1445993-26-9
Color White
Purity Grade Notes Research
Molecular Formula C22H19F2N3O4S
Quantity 50 mg
Synonym ABBV-075
Solubility Information DMSO : 100 mg/mL (217.64 mM; Need ultrasonic)
SMILES CCS(=O)(NC1=CC=C(OC2=CC=C(F)C=C2F)C(C3=CN(C)C(C4=C3C=CN4)=O)=C1)=O
Molecular Weight (g/mol) 459.47
Formula Weight 459.47
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death