missing translation for 'onlineSavingsMsg'
Learn More

Mitotane

Product Code. p-7214323 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16671842

Brand: Thermo Scientific Acros 109850010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53-19-0 | C14H10Cl4 | 320.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-19-0
Molecular Formula C14H10Cl4
Molecular Weight (g/mol) 320.03
InChI Key JWBOIMRXGHLCPP-UHFFFAOYNA-N
IUPAC Name 1-chloro-4-[2,2-dichloro-1-(2-chlorophenyl)ethyl]benzene
SMILES ClC(Cl)C(C1=CC=C(Cl)C=C1)C1=CC=CC=C1Cl

Specifications

Melting Point 77.0°C to 78.0°C
Linear Formula (ClC6H4)2CHCHCl2
Quantity 1 g
Merck Index 15, 6301
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso, methanol, ethanol, isooctane and, ccl4
Formula Weight 320.04
Chemical Name or Material Mitotane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.