Learn More
Motic™ Mirror Set, BA210
Product Code. 15495222
Change view
Click to view available options
For Use With (Equipment):
BA210 Microscope
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15495222 BA210 Microscope Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15495222 Supplier Motic™ Supplier No. 1101001900461
Suitable for use with BA210 Series Upright Compound Microscopes. Motic™ Mirror Set, BA210 focuses light up through the hole in the stage to facilitate specimen viewing.
  • Focus light up through the hole in the microscope stage, or slide platform.
  • Reflect ambient light (especially sunlight) under the slide being viewed to illuminate it.
Specifications
Type Mirror Set
For Use With (Equipment) BA210 Microscope