missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIR96-IN-1, MedChemExpress
Description
MIR96-IN-1 targets the Drosha site in the miR-96 (miRNA-96, microRNA-96) hairpin precursor, inhibiting its biogenesis, derepressing downstream targets, and triggering apoptosis in breast cancer cells. MIR96-IN-1 binds to RNAs with Kds of 1.3, 9.4, 3.4, 1.3 and 7.4 μM for RNA1, RNA2, RNA3, RNA4 and RNA5, respectively.
Specifications
Specifications
| Chemical Name or Material | MIR96-IN-1 |
| CAS | 1311982-88-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C33H48N8O2 |
| Quantity | 1 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (169.84 mM) |
| SMILES | O=C(NCCCN=[N+]=[N-])CCCOC1=C(C(C)(C)C)C=C(C2=NC3=CC=C(N4CCN(C)CC4)C=C3N2)C=C1C(C)(C)C |
| Molecular Weight (g/mol) | 588.79 |
| Formula Weight | 588.79 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?