Learn More
Agilent BioTek Minitube Carrier
Minitube Carrier is for labware up to 50 mm in height, including 1.2 mL minitubes
Brand:  Agilent BioTek 7212042
Additional Details : Weight : 0.50000kg
Product Code. 16562302
  • 545.00 EUR / Each
Estimated Shipment: 05-07-2024
Specifications
1.2 mL Minitubes or Labware up to 50 mm in height
Agilent BioTek Microplate Washer and Reader Instruments