Learn More
Brady™ MiniMark™ High Performance Ribbons
Description
- Excellent solvent resistance, high heat resistance, and super scratch resistance.
- High Performance Resin Ribbon for printing on B-595 Vinyl.
- 110 mm Wide
Specifications
Specifications
| Product Type | Printer Ribbon |
| Color | White |
| For Use With (Equipment) | MiniMark Printer |
| Length (English) | 76.6 yd. |
| Width (English) | 4.33 in. |
| Length (Metric) | 70 m |
| Width (Metric) | 110 mm |
| Diameter (Metric) | 12.7 mm I.D. |
| Dimensions (L x W) | 70 m x 110 mm |
| Description | 110 mm White Ribbon |
| Show More |