Learn More
Lovibond™ Water Testing Minikit
Product Code. 10616151
Click to view available options
Range:
0 to 500 mg/L
0.75 to 10% acid
10 to 500 mg/L
2 to 50 mg/L
20 to 200 mg/L
20 to 500 mg/L
5 to 5000 mg/L
70 to 1500 mg/L
Analytes:
Alkalinity Total M
Alkalinity-P, CaC03
Chloride
Cleaning Acid Strength
Cyanuric Acid
Nitrite
Sulphate (low range) Na2SO4
Sulphite (high range)
Sulphite (low range)
Unit Size:
Each
Product Code. 10616151
Brand: Lovibond™ 414220
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
  • Easy operation and exact reagent dosing
  • High accuracy
  • Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
  • Unrestricted shipment
  • Safe storage
Specifications
Analytes Cyanuric Acid
Includes Flip top case with calibrated two piece tube, tablet reagents and instructions
Range 20 to 200 mg/L
Sufficient For Approximately 40 Tests
Test Method Turbidity
Type Minikit water testing kit
compliance-icons
Product Identifier
  • MINIKIT TEST CYANURIC ACID AF422 .
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : melamine