Learn More
Lovibond™ Water Testing Minikit
Product Code. 10511931
Click to view available options
Range:
0 to 500 mg/L
0.75 to 10% acid
10 to 500 mg/L
2 to 50 mg/L
20 to 200 mg/L
20 to 500 mg/L
5 to 5000 mg/L
70 to 1500 mg/L
Analytes:
Alkalinity Total M
Alkalinity-P, CaC03
Chloride
Cleaning Acid Strength
Cyanuric Acid
Nitrite
Sulphate (low range) Na2SO4
Sulphite (high range)
Sulphite (low range)
Unit Size:
Each
Product Code. 10511931
Brand: Lovibond™ 414340
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
  • Easy operation and exact reagent dosing
  • High accuracy
  • Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
  • Unrestricted shipment
  • Safe storage
Specifications
Analytes Sulphite (low range)
Includes Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions.
Quantity 1 Each
Range 2 to 50 mg/L
Sufficient For Approximately 40 Tests
Test Method Tablet count
Type Minikit water testing kit
compliance-icons
Product Identifier
  • MINIKIT TEST SULFITE LOW AF434
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : sulfamic acid