Learn More
Lovibond™ Water Testing Minikit
Click to view available options
Range:
0 to 500 mg/L
0.75 to 10% acid
10 to 500 mg/L
2 to 50 mg/L
20 to 200 mg/L
20 to 500 mg/L
5 to 5000 mg/L
70 to 1500 mg/L
Analytes:
Alkalinity Total M
Alkalinity-P, CaC03
Chloride
Cleaning Acid Strength
Cyanuric Acid
Nitrite
Sulphate (low range) Na2SO4
Sulphite (high range)
Sulphite (low range)
Unit Size:
Each
Description
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
- Easy operation and exact reagent dosing
- High accuracy
- Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
- Unrestricted shipment
- Safe storage
Specifications
Specifications
| Analytes | Sulphate (low range) Na2SO4 |
| Includes | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. |
| Range | 20 to 200 mg/L |
| Sufficient For | Approximately 40 Tests |
| Test Method | Tablet count |
| Type | Minikit water testing kit |
Safety and Handling
Product Identifier
- MINIKIT TEST SULFATE LOW AF432 .
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Reproductive toxicity Category 2
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H360FD-May damage fertility. May damage the unborn child.
- H361d-Suspected of damaging the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contains : barium chloride dihydrate, salicylic acid, disodium tetraborate, EDTA, boric acid