Learn More
Elma™ Miniature Basket
Description
- Basket for cleaning small parts
- Made of stainless steel
- Compatible with Elma™ Pins
Specifications
Specifications
| Type | Small Basket |
| Diameter (Metric) | 64 mm |
| For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
| Style | 5 Sections |
| Height (Metric) | 16 mm |