Learn More
Bohlender™ Mini1 Basic Desiccator
Click to view available options
Dimensions (L x W):
221 x 214 mm
Unit Size:
Each
Specifications
Specifications
| Dimensions (L x W) | 221 x 214 mm |
| Length (Metric) | 221 mm |
| Width (Metric) | 214 mm |