Learn More
Cytiva Mini Dialysis Kits
Offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.
Brand:  Cytiva 80-6483-75
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10196074
  • 482.00 EUR / 1 set
Estimated Shipment: 21-05-2024
to see stock.
Includes: 50 tubes with 50 dialysis and 50 standard caps, plus six floats
Description
Description
  • Especially well-suited for desalting protein samples prior to analysis by isoelectric focusing (IEF) and 2-D electrophoresis
  • Uses disposable dialysis tubes with dialysis membrane incorporated into caps
  • Conical tube bottom maximizes sample recovery
  • Process complete in as little as several hours
  • Choice of molecular weight cut-off and tube size
Specifications
Specifications
Mini Dialysis Kit
250 μL
Each
1 kDa
50 x tube with dialysis cap, 50 x standard cap, 6 x float
Offers a simple solution to the handling problems of small volume dialysis, the dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane