Learn More
Milnacipran hydrochloride, MedChemExpress
Product Code. 16474267
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16474267
Brand: Medchem Express HYB0168A
MedChemExpress Milnacipran hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the clinical treatment of fibromyalgia.
Milnacipran hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI) used in the clinical treatment of fibromyalgia.
Specifications
Chemical Name or Material Milnacipran hydrochloride
Physical Form Solid
CAS 101152-94-7
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H23ClN2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : ≥ 100 mg/mL (353.59 mM) ∣DMSO : ≥ 48 mg/mL (169.73 mM)
SMILES O=C([C@@]1(C2=CC=CC=C2)[C@@H](CN)C1)N(CC)CC.Cl
Molecular Weight (g/mol) 282.81
Formula Weight 282.81
Percent Purity 99.92%
Grade Research