Learn More
Merck Millicell Hanging Cell Culture Insert
Product Code. 15565960 Shop All Merck Products
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
5 μm
8 μm
No. of Wells:
6
12
24
Unit Size:
Pack of 48
Product Code. 15565960
Brand: Merck MCRP06H48
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Allows attachment or suspension cells to access media from both their apical and basolateral sides.
  • Cell growth, structure, and function more closely mimic what occurs in vivo
  • In addition, Millicell inserts make it possible to study both sides of the cell monolayer
  • The inserts are easily prepared for SEM and TEM visualizing techniques, and they are compatible with cellular and/or fluorescent stains
  • For co-culturing and permeability assays
  • Unique design allows easier basolateral access than other hanging inserts with less risk of contamination
  • Available in 5 pore sizes and 3 diameters, including a 1 μm pore size that is optically transparent for better visualization by microscopy
  • PET Membrane (polyethylene terephthalate) for growth of attachment-dependent cells without matrix
  • Individually blister packaged
TRUSTED_SUSTAINABILITY
Specifications
Description PET (Polyethylene Terephthalate) Filter Code
Material Polystyrene
Pore Size 1 μm
Type Cell Culture Insert
Item Description 50°C Max. Operating Temperature
For Use With (Application) Cell Attachment, Cell Culture, Cell Differentiation, Immunocytochemistry
Diameter (Metric) Filter 24 mm
Quantity 48/Pk
For Use With 6-well Plates
Color Clear
Sterility Sterile
No. of Wells 6