Learn More
Trajan Scientific and Medical™ MicroVolume Syringes
Product Code. 15228483
Click to view available options
Volume (Metric):
5 μL
100 μL
Needle Gauge:
25 G
26 G
Diameter (Metric) Outer Needle:
0.47 mm
0.5 mm
Unit Size:
Each
Product Code. 15228483
Brand: Trajan Scientific and Medical™ 005055
Gives better performance and superior quality. Trajan Scientific and Medical MicroVolume Syringes has the knowledge and expertise that goes into manufacturing an SGE syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Manual injection syringe
Specifications
Description Syringe
Needle Gauge 25 G
Diameter (Metric) Inner Needle 0.20 mm
Diameter (Metric) Outer Needle 0.5 mm
Product Type Removable Needle
Volume (Metric) 100 μL
Length (Metric) Needle 70 mm
Needle Point Style Bevel