Learn More
Ohaus™ Microplate Clamp
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
Microplates
Unit Size:
Each
Description
- OHAUS offers a range of shaker accessories to meet specific sample requirements
- A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
- Platforms can increase the amount of usable space for samples
- In some cases can be stacked to increase throughput and double sample capacity
- Tube racks and dimpled mats securely hold tubes in place
Specifications
Specifications
| Type | Clamp |
| Material | Stainless Steel |
| For Use With (Equipment) | Microplates |
| Description | Clamp Microplate Stainless Steel |