Learn More
Elga™ Micromeg Disposable Replacement Cartridge
Product Code. 13135940
Change view
Click to view available options
Flow Rate:
90 L/hr.
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
13135940 90 L/hr. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13135940 Supplier Elga™ Supplier No. COLDILP1015
For the production of low volumes of deionised water.
  • Produces water up to a quality of 10MΩ/cm at flows of up to 90L/hr
  • Requires no electrical input
  • Cartridges can be replaced quickly
Specifications
Flow Rate 90 L/hr.
Filter Size MC:DW cartridges
For Use With (Equipment) Micromeg Disposable Deionizer PRDIDC0020
Inlet Pressure 3.4 bar