missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micrococcus Lysodeikticus, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
From Micrococcus lysodeikticus
Specifications
Specifications
| Content And Storage | 4°C |
| Form | Lyophilized |
| Product Type | Micrococcus lysodeikticus |
| For Use With (Application) | Assay of lysozyme |
| Quantity | 1 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction