missing translation for 'onlineSavingsMsg'
Learn More

Micrococcus Lysodeikticus, MP Biomedicals

Product Code. 11460142 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11460142

Brand: MP Biomedicals 0215997201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lyophilized cells suitable as a lysozyme substrate.

From Micrococcus lysodeikticus

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Lyophilized
Product Type Micrococcus lysodeikticus
For Use With (Application) Assay of lysozyme
Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.