Learn More
SI Analytics™ Micro Ubbelohde Viscometer
Product Code. 10159990 Shop All SI Analytics Products
Click to view available options
Viscosity Range:
0.4 to 6 cSt
12 to 180 cSt
4 to 60 cSt
40 to 800 cSt
Approximate Constant:
0.01K
0.1K
0.3K
1K
Size:
0.4mm Diameter
0.70mm Diameter
0.95mm Diameter
1.26mm Diameter
Unit Size:
Each
Product Code. 10159990
Brand: SI Analytics™ 285401136
Only null left
Add to basket
Add to basket
Determine the limit viscosity number of polymers. SI Analytics™ Micro Ubbelohde Viscometer includes ring marks to ensure that they can be checked by means of manual measurements.
  • Suitable for transparent liquids manual and automatic measurement, foaming liquids, liquid mixture with highly volatile components, high or low temperature measurements, and minimum measurement substance or rinsing agent quantities
  • The recommended minimum flow through time is 200 seconds
  • Calibrated
  • With constant for automatic measurements
  • Filling quantity of 3 to 4mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viscometer
Viscosity Range 12 to 180 cSt
Calibrated Yes
Approximate Constant 0.3K
Certifications/Compliance DIN 55 350, Part 18 and DIN 51562, Part 2
Includes Ring Marks
For Use With (Application) For Determination of Absolute and Relative, Kinematic Viscosity of Liquids With Newtonian Flow Behavior
Size 0.95mm Diameter
Volume (Metric) 3 to 4 mL