Learn More
Fisherbrand™ Micro Insert for well plates, glass
Product Code. 10295072 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
400 μL
1000 μL
1200 μL
Style:
Constricted
Round
Tapered
Unit Size:
Pack of 960
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Style unitSize
10295072 400 μL Tapered Pack of 960
10091762 1000 μL Constricted Pack of 960
10275772 1200 μL Round Pack of 960
10377552 1000 μL Round Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10295072
Only null left
Add to basket
Add to basket
For complex UHPLC, LC and GC applications.
  • Clear
  • Glass
  • Plates meet ANSI and SBS standards
  • Different sealing options
  • Chromatography tested
  • No leeching due to high-end material
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Glass
Volume (Metric) 400 μL
Style Tapered