missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MHC Class II Monoclonal Antibody (HIS19), eBioscience™, Invitrogen™

Product Code. p-7218764
Change view
Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17120532 2 mg 2mg
17130532 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17120532 Supplier Invitrogen™ Supplier No. 14092037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Description: The HIS19 monoclonal antibody reacts with a polymorphic epitope of the rat MHC class II molecule, RT1B. This antibody reacts with all rat haplotypes except the RT1n (including BN and MAXX strains). The rat MHC class II (I-A equivalent), RT1B, is expressed by antigen presenting cells including B cells, dendritic cells and macrophages. Applications Reported: The HIS19 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining of frozen tissue sections. Applications Tested: The HIS19 antibody has been tested by flow cytometric analysis of rat spleen or lymph node cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

MHC (major histocompatibility complex) class II molecules are transmembrane glycoproteins expressed on the surface of professional antigen-presenting cells, such as macrophages, dendritic cells and B cells. Before their exposition on the cell surface, the MHC class II molecules react with endocytosed exogenous antigens, which are then presented to the T cells. The antigen-binding grove between MHC class II alpha and beta chain is open at both ends and is 15-24 amino acid residues long. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class II
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone HIS19
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias beta 1 domain; B-L; B-L beta chain; B-L beta minor; B-LB; BLB major; BLB1; BLB2; B-LB2; B-LB21; B-LB21 major; BLB3; Bl-Beta II protein; B-LBII; class II histocompatibility antigen, B-L beta chain; class II histocompatibility antigen, M alpha chain; D6S222E; DMA; FECA-DRB; GSP-BLB1; GSP-BLB2; HLA class II histocompatibility antigen, DM alpha chain; HLADM; HLA-DMA; major histocompatibility class II antigen B-L beta; major histocompatibility complex class II B; Major histocompatibility complex class II beta chain BLB1; Major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); major histocompatibility complex class II beta chain BLB1, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB2, (similar to HLA class II, D beta chain); Major histocompatibility complex class II beta chain BLB3, (similar to HLA class II, D beta chain); major histocompatibility complex, class II, DM alpha; MHC 2; MHC Class II; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen B-F minor heavy chain; MHC class II antigen DMA; MHC class II antigen DRB; MHC Class II beta 1 and 2 domains; MHC class II beta 1 domain; MHC class II beta chain; MHC class II beta chain 1; MHC class II beta chain 2; MHC class II B-L beta chain; MHC II; MHCCII; MHCFECA-DRB; really interesting new gene 6 protein; RING6; S19; uncharacterized protein LOC101747454
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.