missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MgcRacGAP/RACGAP1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47267JF646
This item is not returnable.
View return policy
Description
MgcRacGAP/RACGAP1 Polyclonal antibody specifically detects MgcRacGAP/RACGAP1 in Human samples. It is validated for Western Blot
Specifications
| MgcRacGAP/RACGAP1 | |
| Polyclonal | |
| Western Blot | |
| GTPase activating protein, HsCYK-4, ID-GAP, KIAA1478, Male germ cell RacGap, MGCRACGAP, MgcRacGAPrac GTPase-activating protein 1, Rac GTPase activating protein 1 | |
| The immunogen for this product maps to a region between residue 150 and 200 of human Rac GTPase-Activating Protein 1 using the numbering given in entry NP_001119575.1 (GeneID 29127). | |
| 0.1 mL | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 29127 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction