Learn More
Metro™ Super Erecta™ Wire Shelf, Chrome Finish
Click to view available options
Dimensions (L x W):
24 x 18 in. (61 x 45.7 cm)
24 x 21 in. (61 x 53 cm)
30 x 21 in. (76 x 53 cm)
30 x 24 in. (76 x 61 cm)
36 x 18 in. (91.4 x 45.7 cm)
36 x 21 in. (91.4 x 53 cm)
36 x 24 in. (91.4 x 61 cm)
36 x 30 in. (91.4 x 76 cm)
36 x 36 in. (91 x 91 cm)
42 x 14 in. (106.6 x 35.5 cm)
42 x 21 in. (106.6 x 53 cm)
42 x 24 in. (106.6 x 61 cm)
48 x 18 in. (121.9 x 45.7 cm)
48 x 21 in. (122 x 53 cm)
48 x 36 in. (121.9 x 91 cm)
60 x 14 in. (152.4 x 35.5 cm)
60 x 18 in. (152.4 x 45.7 cm)
60 x 21 in. (152.4 x 53 cm)
60 x 24 in. (152.4 x 61 cm)
60 x 30 in. (152.4 x 76 cm)
72 x 21 in. (183 x 53 cm)
72 x 30 in. (183 x 76 cm)
72 x 36 in. (183 x 91 cm)
Load Bearing Capacity (English):
21 lb.
400 lb.
600 lb.
800 lb.
Unit Size:
Each
Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Real nickel-chrome finish for dry, low humidity environment
- Protective lacquer coated
Specifications
Specifications
| Product Type | Wire Shelf |
| Dimensions (L x W) | 36 x 21 in. (91.4 x 53 cm) |
| For Use With (Equipment) | Metro™ Super Erecta™ shelving system |
| Includes | Plastic sleeves and wedges |
| Length (English) | 36 in. |
| Length (Metric) | 91.4 cm |
| Load Bearing Capacity (English) | 800 lb. |
| Load Bearing Capacity (Metric) | 363 kg |
| Width (English) | 21 in. |
| Width (Metric) | 53 cm |
| Show More |