Learn More
Metro™ Standard-Duty Mobile Security Units, Stainless Steel
Product Code. 15957231
Click to view available options
Width (English):
21.5 in.
27.25 in.
33.5 in.
Length (English):
40.75 in.
52.75 in.
65 in.
Weight (English):
138 lb.
154 lb.
157 lb.
167 lb.
174 lb.
193 lb.
195 lb.
215 lb.
Unit Size:
Each
Product Code. 15957231
Brand: Metro™ SEC33SSD
Only null left
Add to basket
Add to basket
Standard-duty mobile security units provide protection of valuable materials and sensitive items from loss or pilferage. Available in a sleek, easy-to-clean stainless steel finish.
  • Mobile security units provide protection for valuables and sensitive materials from loss or pilferage.
  • Type 304 Stainless provides superior corrosion resistance.
  • Heavy gauge open wire construction keeps contents visible at all times, making it easy to check inventory.
  • Choice of casters to match requirements.
  • Patented, ergonomic, 1/4-turn door handle.
  • Double doors open 270° and can be secured along the sides of the unit.
  • Fixed top and bottom Super Erecta shelves.
  • Optional intermediate shelves can be postitioned in 1 in. increments along entire height of post.
Order Info
Shipped knocked down; assemble easily. Super Adjustable/Super Erecta™ intermediate shelves and casters sold separately.
Specifications
Product Type Mobile Security Cart
Capacity (Metric) 63 kg
Color Silver
Dimensions (L x W x H) 40.75 x 21.5 x 62 in. (103.5 x 54.6 x 157.5 cm)
Height (English) 62 in.
Height (Metric) 157.5 cm
Length (English) 40.75 in.
Length (Metric) 103.5 cm
Material Stainless Steel
No. of Shelves 2
Weight (Metric) 63 kg
Weight (English) 138 lb.
Width (English) 21.5 in.
Width (Metric) 54.6 cm