Learn More
Emma Safety Footwear Metric Safety Shoes
Product Code. 15928541
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15928541 35 1 pair
15938541 36 1 pair
15948541 37 1 pair
15958541 38 1 pair
15968541 39 1 pair
15978541 40 1 pair
15988541 41 1 pair
15998541 42 1 pair
15908551 43 1 pair
15918551 44 1 pair
15928551 45 1 pair
15938551 46 1 pair
15948551 47 1 pair
15958551 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15928541 Supplier Emma Safety Footwear Supplier No. MM50654035
Only null left
Add to basket
Add to basket
Unique microfiber upper is fully washable. Features SmartecAir™ for dry, fresh feet. Protective toe cap prevents staining and is lubricant and fat resistant. The sole was developed for use on hard tiled floors and other easy-to-clean floors often seen in high-tech and cleanroom environments.
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Easy Twist Outsole
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Elastic
Color White
Design Metric
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 35
Toe Material Steel