Learn More
Methylhexanamine.HCl, Lipomed
Description
Methylhexanamine.HCl
Specifications
Specifications
| Chemical Name or Material | Methylhexanamine hydrochloride |
| CAS | 13803-74-2 |
| Analyte or Component Names | Methylhexanamine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H17 N . Cl H |
| Synonym | Dimethylamylamine Hydrochloride, 4-Methyl-2-hexanamine Hydrochloride, DMAA HCl |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CCC(C)CC(C)N |
| IUPAC Name | 4-methylhexan-2-amine;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |