Learn More
(Methylamino)acetaldehyde dimethyl acetal, 97%
Product Code. 11367685
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367685 25 g 25g
11377685 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11367685 Supplier Thermo Scientific Alfa Aesar Supplier No. L02391.14
Request Bulk or Custom Format
CAS: 122-07-6 | C5H13NO2 | 119.164 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 122-07-6
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.164
MDL Number MFCD00008485
InChI Key HUMIEJNVCICTPJ-UHFFFAOYSA-N
Synonym methylaminoacetaldehyde dimethyl acetal, methylamino acetaldehyde dimethyl acetal, ethanamine, 2,2-dimethoxy-n-methyl, 2,2-dimethoxyethyl methyl amine, n-methylaminoacetaldehyde dimethyl acetal, n-2,2-dimethoxyethyl methylamine, unii-6v8q54khcl, 2,2-dimethoxyethylmethylamine, 2-methylamino acetaldehyde dimethyl acetal
PubChem CID 8503
IUPAC Name 2,2-dimethoxy-N-methylethanamine
SMILES CNCC(OC)OC
Specifications
Melting Point -73°C
Density 0.928
pH 10.9
Boiling Point 139°C to 140°C
Flash Point 29°C (84°F)
Odor Ammonia-like
Refractive Index 1.4115
Quantity 25 g
UN Number UN1993
Beilstein 605322
Sensitivity Air Sensitive
Formula Weight 119.16
Percent Purity 97%
Chemical Name or Material (Methylamino)acetaldehyde dimethyl acetal