Learn More
Methylamine hydrochloride, 99%
Product Code. 10562171 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
500 g
1 kg
5 kg
Unit Size:
1kg
250g
500g
50g
5kg
Product Code. 10562171
Brand: Thermo Scientific Acros 156780010
Request Bulk or Custom Format
CAS: 593-51-1 | CH6ClN | 67.52 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 593-51-1
Molecular Formula CH6ClN
Molecular Weight (g/mol) 67.52
InChI Key NQMRYBIKMRVZLB-UHFFFAOYSA-N
IUPAC Name methanaminium chloride
SMILES [Cl-].C[NH3+]
Specifications
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3NH2·HCl
Quantity 1 kg
Formula Weight 67.52
Chemical Name or Material Methylamine hydrochloride