missing translation for 'onlineSavingsMsg'
Learn More

Methyl deacetylasperulosidate, MedChemExpress

Product Code. p-200101357
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16463926

Brand: Medchem Express HYN1503

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methyl deacetylasperulosidate is an iridoid and shows purgative effects in mice and lowers the blood glucose level in normal mice.

Methyl deacetylasperulosidate is an iridoid and shows purgative effects in mice and lowers the blood glucose level in normal mice.

Specifications

Chemical Name or Material Methyl deacetylasperulosidate
CAS 52613-28-2
Color White
Purity Grade Notes Research
Molecular Formula C17H24O11
Quantity 25 mg
Synonym 6α-Hydroxygeniposide Deacetylasperulosidic acid methyl ester
Solubility Information DMSO : 250 mg/mL (618.25 mM; Need ultrasonic)
SMILES O=C(C1=CO[C@@H](O[C@H]2[C@@H]([C@H]([C@@H]([C@@H](CO)O2)O)O)O)[C@@]3([H])[C@]1([H])[C@@H](O)C=C3CO)OC
Molecular Weight (g/mol) 404.37
Formula Weight 404.37
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.