missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyclopentanecarboxylate, 97%

Product Code. p-7043428
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11340194

Brand: Thermo Scientific Alfa Aesar L00741.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4630-80-2 | C7H12O2 | 128.17 g/mol

Methyl cyclopentanecarboxylate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cyclopentanecarboxylate

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4630-80-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00019290
InChI Key IIHIJFJSXPDTNO-UHFFFAOYSA-N
Synonym cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl
PubChem CID 78365
IUPAC Name methyl cyclopentanecarboxylate
SMILES COC(=O)C1CCCC1

Specifications

Boiling Point 81°C to 82°C (5 mmHg)
Refractive Index 1.436
Quantity 5 g
Beilstein 2040762
Solubility Information Difficult to mix.
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Methyl cyclopentanecarboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.