Learn More
Methyl cyclopentanecarboxylate, 97%
Product Code. 11340194
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340194 5 g 5g
11350194 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11340194 Supplier Thermo Scientific Alfa Aesar Supplier No. L00741.06
Request Bulk or Custom Format
CAS: 4630-80-2 | C7H12O2 | 128.17 g/mol
Methyl cyclopentanecarboxylate
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl cyclopentanecarboxylate
Solubility
Difficult to mix.
Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4630-80-2
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00019290
InChI Key IIHIJFJSXPDTNO-UHFFFAOYSA-N
Synonym cyclopentanecarboxylic acid, methyl ester, cyclopentanecarboxylic acid methyl ester, methyl cyclopentanoate, acmc-1am5n, methylcyclopentanecarboxylate, methylcyclopentane carboxylate, methyl cyclopentane carboxylate, methyl-1-cyclopentanecarboxylate, cyclopentanoic acid methyl ester, cyclopentanecarboxylic acid methyl
PubChem CID 78365
IUPAC Name methyl cyclopentanecarboxylate
SMILES COC(=O)C1CCCC1
Specifications
Boiling Point 81°C to 82°C (5 mmHg)
Refractive Index 1.436
Quantity 5 g
Beilstein 2040762
Solubility Information Difficult to mix.
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material Methyl cyclopentanecarboxylate