missing translation for 'onlineSavingsMsg'
Learn More

Methyl Beta-D-Glucuronide, Sodium Salt, TRC

Product Code. 30133997
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133997 1 g 1g
30011877 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133997 Supplier Toronto Research Chemicals Supplier No. TRCM3115501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134253-42-2
Molecular Formula C7 H11 O7 . Na
Molecular Weight (g/mol) 230.15
Synonym 1-O-Methyl-B-D-Glucuronic Acid Sodium Salt
IUPAC Name sodium;(2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methoxyoxane-2-carboxylate
SMILES [Na+].CO[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H12O7.Na/c1-13-7-4(10)2(8)3(9)5(14-7)6(11)12;/h2-5,7-10H,1H3,(H,11,12);/q;+1/p-1/t2-,3-,4+,5-,7+;/m0./s1
Formula Weight 230.0402
Chemical Name or Material Methyl beta-D-Glucuronide, Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.