missing translation for 'onlineSavingsMsg'
Learn More

Methyl (3R)-3-[tert-Butyl(dimethyl)silyl]oxy-5-oxo-6-(triphenylphosphanylidene)hexanoate, Mikromol™

Product Code. 30459563
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459563 4 x 25 mg 100mg
30049682 25 mg 25mg
30050508 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30459563 Supplier Mikromol Supplier No. MM0936.0800254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl (3R)-3-[tert-Butyl(dimethyl)silyl]oxy-5-oxo-6-(triphenylphosphanylidene)hexanoate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (3R)-3-[tert-Butyl(dimethyl)silyl]oxy-5-oxo-6-(triphenylphosphanylidene)hexanoate
CAS 147118-35-2
Analyte or Component Names Methyl (3R)-3-[tert-Butyl(dimethyl)silyl]oxy-5-oxo-6-(triphenylphosphanylidene)hexanoate
Shipping Condition Room Temperature
Molecular Formula C31 H39 O4 P Si
Certifications/Compliance ISO 17025
Synonym Hexanoic acid, 3-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-5-oxo-6-(triphenylphosphoranylidene)-, methyl ester, (R)-, (3R)-3-(tert-Butyldimethylsilanyloxy)-5-oxo-6-(triphenylphosphanylidene)hexanoic acid methyl ester, (R)-3-[(tert-Butyldimethylsilanyl)oxy]-5-oxo-6-(triphenylphosphanylidene)hexanoic acid methyl ester, R-(-)-Oxo-3-(tert-butyldimethylsilyloxy)-6-triphenylphosphoranediylhexanoic acid methyl ester, Hexanoic acid, 3-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-5-oxo-6-(triphenylphosphoranylidene)-, methyl ester, (3R)-, Methyl (3R)-3-[tert-butyl(dimethyl)silyl]oxy-5-oxo-6-(triphenyl-$l^{5}-phosphanylidene)hexanoate
InChI Formula InChI=1S/C31H39O4PSi/c1-31(2,3)37(5,6)35-26(23-30(33)34-4)22-25(32)24-36(27-16-10-7-11-17-27,28-18-12-8-13-19-28)29-20-14-9-15-21-29/h7-21,24,26H,22-23H2,1-6H3/t26-/m1/s1
Solution Type Neat
SMILES COC(=O)C[C@@H](CC(=O)C=P(c1ccccc1)(c2ccccc2)c3ccccc3)O[Si](C)(C)C(C)(C)C
IUPAC Name methyl (3R)-3-[tert-butyl(dimethyl)silyl]oxy-5-oxo-6-(triphenyl-λ^{5}-phosphanylidene)hexanoate
Molecular Weight (g/mol) 534.7
Quantity 4 x 25 mg
Formula Weight 534.2355 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.