missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-nitrobenzoate, 98+%

Product Code. p-7022397
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11482138

Brand: Thermo Scientific Alfa Aesar A15155.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 618-95-1 | C8H7NO4 | 181.147 g/mol

Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-nitrobenzoate was used in the preparation of iodoarene.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-95-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007250
InChI Key AXLYJLKKPUICKV-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
PubChem CID 69260
IUPAC Name methyl 3-nitrobenzoate
SMILES COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 77°C to 80°C
Boiling Point 279°C
Quantity 50 g
Beilstein 392449
Solubility Information Insoluble in water.
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material Methyl 3-nitrobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.