missing translation for 'onlineSavingsMsg'
Learn More

Methyl-[3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]carbamic Acid 1,1-Dimethylethyl Ester, TRC

Product Code. 30481293
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481293 50 mg 50mg
30476608 10 mg 10mg
30464802 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481293 Supplier Toronto Research Chemicals Supplier No. TRCM32079050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl-[3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]carbamic Acid 1,1-Dimethylethyl Ester
Analyte or Component Names Methyl-[3-(1-naphthalenyloxy)-3-(2-thienyl)propyl]carbamic Acid 1,1-Dimethylethyl Ester
Shipping Condition Room Temperature
Molecular Formula C23 H27 N O3 S
InChI Formula InChI=1S/C23H27NO3S/c1-23(2,3)27-22(25)24(4)14-13-20(21-10-7-15-28-21)26-19-12-11-17-8-5-6-9-18(17)16-19/h5-12,15-16,20H,13-14H2,1-4H3
Solution Type Neat
SMILES CN(CCC(Oc1ccc2ccccc2c1)c3cccs3)C(=O)OC(C)(C)C
IUPAC Name tert-butyl N-methyl-N-(3-naphthalen-2-yloxy-3-thiophen-2-ylpropyl)carbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 397.53
Quantity 50 mg
Formula Weight 397.171
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.