missing translation for 'onlineSavingsMsg'
Learn More

Methoxyphenamine Hydrochloride, MP Biomedicals™

Product Code. 11407722 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11407722

Brand: MP Biomedicals 0215542901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Soluble in water (50mg/mL)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-3-5588
Molecular Formula C11H18ClNO
Molecular Weight (g/mol) 215.72
MDL Number MFCD00034832
InChI Key WJVCROKJTIUCAZ-UHFFFAOYNA-N

Specifications

Color White
Quantity 1 g
Synonym methoxyphenamine hydrochloride, methoxyphenamine hcl, orthoxine hydrochloride, ortodrinex hydrochloride, methoxyphenaminium chloride, methoxiphenadrin hydrochloride, o-methoxymethamphetamine hydrochloride, 1-2-methoxyphenyl propan-2-yl methyl amine hydrochloride, 1-2-methoxyphenyl-n-methylpropan-2-amine hydrochloride
SMILES CC(CC1=CC=CC=C1OC)NC.Cl
IUPAC Name 1-(2-methoxyphenyl)-N-methylpropan-2-amine;hydrochloride
PubChem CID 9859211
Formula Weight 215.7
Physical Form Powder
Chemical Name or Material Methoxyphenamine Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.