missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate, MP Biomedicals™

Product Code. 11463530 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11463530

Brand: MP Biomedicals 0210229991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acts as a potent anti-cancer agent. Methotrexate, MP Biomedicals is a folic acid antagonist that is structurally similar to folic acid. It is a mixture of 4-amino-10-methylfolic acid and related compounds. It is a potent inhibitor of dihydrofolate reducta

Blocks DNA synthesis by blocking the production of tetrahydrofolate cofactors required for the synthesis of thymidine. Methotrexate is actively transported into cells by the folate transporter. In the cell, it is converted to a high molecular weight polyglutamate metabolite by folypolyglutamate synthase that, in turn, binds to dihydrofolate reductase and inhibits its activity. Methotrexate-polyglutamate is degraded intracellularly by gamma-glutamyl hydrolase.

Methotrexate is an allosteric inhibititor of dihydrofolate reductase (DHFR), the enzyme that catalyzes the conversion of dihydrofolate to tetrahydrofolate. Since tetrahydrolfolate is required for purine and pyrimidine synthesis, methotrexate treatment results in the inhibition of DNA and RNA synthesis.

  • A potent inhibitor of dihydrofolate reductase and is an agent for antitumor studies
  • Used to inhibit dihydrofolate reductase in DHFR-based protein expression systems
  • Effective in treatment of pyrimethamine-resistant Plasmodium vivax malaria parasites
  • Also shows immunosuppressive effects, e.g., in rheumatoid arthritis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-05-2
Molecular Formula C20H22N8O5
Molecular Weight (g/mol) 454.447
InChI Key FBOZXECLQNJBKD-ZDUSSCGKSA-N
Synonym methotrexate, abitrexate, rheumatrex, amethopterin, mexate, metatrexan, hdmtx, trexall, methylaminopterinum, antifolan
PubChem CID 126941
ChEBI CHEBI:44185
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
SMILES CN(CC1=CN=C2C(=N1)C(=NC(=N2)N)N)C3=CC=C(C=C3)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 185°C to 204°C
Boiling Point 184°C to 204°C (monohydrate decomposition)
Quantity 1 g
Solubility Information Soluble in alkaline solutions with decomposition. Insoluble in water, ethanol, chloroform and ether.
Optical Rotation 19–24°
Formula Weight 454.4
Grade USP
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.